Merck
NMR spectroscopic analysis of new spiro-piperidylrifamycins.
Magnetic resonance in chemistry : MRC (2005-01-28)
Eduardo Rubio, Isabel Merino, Ana-Belén García, María-Paz Cabal, Cristina Ribas, Miguel Bayod-Jasanada
ABSTRACT
New spiro-piperidylrifamycin derivatives are presented. These compounds were synthesized from the reaction of 3-amino-4-iminorifamycin S and enantiomerically pure 4-piperidones, which generate diasteroisomeric rifabutin analogues with a new stereocentre at the spiranic carbon. The (1)H and (13)C NMR spectra of these new compounds, and also the configuration of the new stereogenic centres, were assigned using 2D NMR spectroscopic techniques. A preliminary study of the (1)H and (13)C NMR spectra of the starting compounds rifamycin S, 3-amino-4-iminorifamycin S and the related rifabutin was also carried out and as a result, their previously published (13)C NMR data were corrected.