Merck
  • Directed control of electroosmotic flow in nonaqueous electrolytes using poly(ethylene glycol) coated capillaries.
Directed control of electroosmotic flow in nonaqueous electrolytes using poly(ethylene glycol) coated capillaries.
Electrophoresis (2001-04-12)
D Belder, H Husmann, J Warnke
ABSTRACT
Poly(ethylene glycol) (PEG)-coated capillaries exhibit unique properties in nonaqueous electrolytes. Immobilized PEG interacts significantly with different cations present in nonaqueous electrolytes. This can induce a positive surface charge on PEG-coated capillaries and results in an adjustable anodic electroosmotic flow (EOF) in nonaqueous electrolytes whereas a reduced cathodic EOF is observed in aqueous electrolytes. The EOF can reversibly be adjusted by the variation of the electrolyte constitution, namely the type of the solvent used and the nature and concentration of background cations. In methanol and especially in acetonitrile electrolytes the magnitude and also the direction of EOF is strongly dependent on the water content. Using different alkali metal cations, the EOF can be increased, reduced, or even reversed depending on the nature of the cation. The directed manipulation of EOF in methanolic electrolytes using PEG-coated capillaries was applied for optimization of nonaqueous capillary electrophoretic separations of acidic compounds with regard to reproducibility, resolution, and analysis time.