Merck
  • Autologous transplantation as consolidation for aggressive non-Hodgkin's lymphoma.
Autologous transplantation as consolidation for aggressive non-Hodgkin's lymphoma.
The New England journal of medicine (2013-11-01)
Patrick J Stiff, Joseph M Unger, James R Cook, Louis S Constine, Stephen Couban, Douglas A Stewart, Thomas C Shea, Pierluigi Porcu, Jane N Winter, Brad S Kahl, Thomas P Miller, Raymond R Tubbs, Deborah Marcellus, Jonathan W Friedberg, Kevin P Barton, Glenn M Mills, Michael LeBlanc, Lisa M Rimsza, Stephen J Forman, Richard I Fisher
ABSTRACT
The efficacy of autologous stem-cell transplantation during the first remission in patients with diffuse, aggressive non-Hodgkin's lymphoma classified as high-intermediate risk or high risk on the International Prognostic Index remains controversial and is untested in the rituximab era. We treated 397 patients who had disease with an age-adjusted classification of high risk or high-intermediate risk with five cycles of cyclophosphamide, doxorubicin, vincristine, and prednisone (CHOP) or CHOP plus rituximab. Patients with a response were randomly assigned to receive three additional cycles of induction chemotherapy (control group) or one additional cycle of induction chemotherapy followed by autologous stem-cell transplantation (transplantation group). The primary efficacy end points were 2-year progression-free survival and overall survival. Of 370 induction-eligible patients, 253 were randomly assigned to the transplantation group (125) or the control group (128). Forty-six patients in the transplantation group and 68 in the control group had disease progression or died, with 2-year progression-free survival rates of 69 and 55%, respectively (hazard ratio in the control group vs. the transplantation group, 1.72; 95% confidence interval [CI], 1.18 to 2.51; P=0.005). Thirty-seven patients in the transplantation group and 47 in the control group died, with 2-year overall survival rates of 74 and 71%, respectively (hazard ratio, 1.26; 95% CI, 0.82 to 1.94; P=0.30). Exploratory analyses showed a differential treatment effect according to risk level for both progression-free survival (P=0.04 for interaction) and overall survival (P=0.01 for interaction). Among high-risk patients, the 2-year overall survival rate was 82% in the transplantation group and 64% in the control group. Early autologous stem-cell transplantation improved progression-free survival among patients with high-intermediate-risk or high-risk disease who had a response to induction therapy. Overall survival after transplantation was not improved, probably because of the effectiveness of salvage transplantation. (Funded by the National Cancer Institute, Department of Health and Human Services, and others; SWOG-9704 ClinicalTrials.gov number, NCT00004031.).
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard