Merck
  • Correlation between the area of high-signal intensity on SPIO-enhanced MR imaging and the pathologic size of sentinel node metastases in breast cancer patients with positive sentinel nodes.
Correlation between the area of high-signal intensity on SPIO-enhanced MR imaging and the pathologic size of sentinel node metastases in breast cancer patients with positive sentinel nodes.
BMC medical imaging (2013-09-14)
Kazuyoshi Motomura, Tetsuta Izumi, Souichirou Tateishi, Hiroshi Sumino, Atsushi Noguchi, Takashi Horinouchi, Katsuyuki Nakanishi
ABSTRACT
We previously demonstrated that superparamagnetic iron oxide (SPIO)-enhanced MR imaging is promising for the detection of metastases in sentinel nodes localized by CT-lymphography in patients with breast cancer. The purpose of this study was to determine the predictive criteria of the size of nodal metastases with SPIO-enhanced MR imaging in breast cancer, with histopathologic findings as reference standard. This study included 150 patients with breast cancer. The patterns of SPIO uptake for positive sentinel nodes were classified into three; uniform high-signal intensity, partial high-signal intensity involving ≥50% of the node, and partial high-signal intensity involving <50% of the node. Imaging results were correlated with histopathologic findings. Thirty-three pathologically positive sentinel nodes from 30 patients were evaluated. High-signal intensity patterns that were uniform or involved ≥50% of the node were observed in 23 nodes that contained macro-metastases and no node that contained micro-metastases, while high-signal intensity patterns involving <50% of the node were observed in 2 nodes that contained macro-metastases and 8 nodes that contained micro-metastases. When the area of high-signal intensity was compared with the pathological size of the metastases, a pathologic >2 mm sentinel node metastases correlated with the area of high-signal intensity, however, a pathologic ≤2 mm sentinel node metastases did not. High-signal intensity patterns that are uniform or involve ≥50% of the node are features of nodes with macro-metastases. The area of high-signal intensity correlated with the pathological size of metastases for nodes with metastases >2 mm in this series.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 670,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 50,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 150,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran, enzymatic synth.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 25,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 410,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 5,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 12,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextran, analytical standard, for GPC, 270,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000