Merck
  • Improving colorectal cancer screening in family medicine: an Oklahoma Physicians Resource/Research Network (OKPRN) study.
Improving colorectal cancer screening in family medicine: an Oklahoma Physicians Resource/Research Network (OKPRN) study.
Journal of the American Board of Family Medicine : JABFM (2013-09-06)
Dewey C Scheid, Robert M Hamm, Kalyanakrishnan Ramakrishnan, Laine H McCarthy, James W Mold
ABSTRACT
The purpose of this study was to describe colorectal cancer screening (CRCS) practices across a variety of primary care clinics and identify the methods used by primary care physicians (PCPs) with higher rates of CRCS ("exemplars"). Physician questionnaires, structured interviews, medical record abstractions, and practice observations were conducted for 48 PCPs in 25 practices within a regional practice-based research network followed by secondary in-depth interviews to further investigate the practices of PCPs in the top quartile of CRCS rates ("exemplars"). We abstracted 3596 medical records (mean of 75 records per PCP). Overall, exemplars had higher CRCS rates (median, 57.2% vs. 27.6%; P < .001). Patients of exemplars had higher screening rates for fecal occult blood testing (FOBT) and colonoscopy but not for flexible sigmoidoscopy or double-contrast barium enemas. Exemplars adopted few of the system-based innovations proposed by researchers to improve CRCS. Colonoscopy was promoted as the preferred CRCS method. FOBT was recommended for patients who could not afford or did not want colonoscopy. Flexible sigmoidoscopy or barium enemas were rarely recommended. Exemplars used brief CRCS promotion scripts that informally paralleled theory-driven counseling techniques. Experienced PCPs use brief CRCS promotion scripts including counseling techniques that improve CRCS performance. Future research should be directed toward whether these techniques can be used to create an intervention aimed at PCPs to improve CRCS.
Sigma-Aldrich
Barium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Barium sulfate, meets USP testing specifications
Sigma-Aldrich
Barium sulfate, LR, ≥98%
Sigma-Aldrich
Barium sulfate, ReagentPlus®, 99%