Merck
Adhesion force between cyclopentane hydrate and mineral surfaces.
Langmuir : the ACS journal of surfaces and colloids (2013-11-26)
Zachary M Aman, William J Leith, Giovanny A Grasso, E Dendy Sloan, Amadeu K Sum, Carolyn A Koh
ABSTRACT
Clathrate hydrate adhesion forces play a critical role in describing aggregation and deposition behavior in conventional energy production and transportation. This manuscript uses a unique micromechanical force apparatus to measure the adhesion force between cyclopentane hydrate and heterogeneous quartz and calcite substrates. The latter substrates represent models for coproduced sand and scale often present during conventional energy production and transportation. Micromechanical adhesion force data indicate that clathrate hydrate adhesive forces are 5-10× larger for calcite and quartz minerals than stainless steel. Adhesive forces further increased by 3-15× when increasing surface contact time from 10 to 30 s. In some cases, liquid water from within the hydrate shell contacted the mineral surface and rapidly converted to clathrate hydrate. Further measurements on mineral surfaces with physical control of surface roughness showed a nonlinear dependence of water wetting angle on surface roughness. Existing adhesive force theory correctly predicted the dependence of clathrate hydrate adhesive force on calcite wettability, but did not accurately capture the dependence on quartz wettability. This comparison suggests that the substrate surface may not be inert, and may contribute positively to the strength of the capillary bridge formed between hydrate particles and solid surfaces.
Sigma-Aldrich
SODOSIL RAM 05, acid cleaner with formic acid
Sigma-Aldrich
SODOSIL RA 14, mildly alkaline, chlorine and phosphate-free cleaner
Sigma-Aldrich
SODOSIL RA 16, alkaline cleaner with chlorine