Merck
  • Efficacy and safety of levodropropizine and dihydrocodeine on nonproductive cough in primary and metastatic lung cancer.
Efficacy and safety of levodropropizine and dihydrocodeine on nonproductive cough in primary and metastatic lung cancer.
The European respiratory journal (1998-08-13)
G Luporini, S Barni, E Marchi, L Daffonchio
ABSTRACT
Nonproductive cough is a frequent and distressing symptom in patients with lung cancer, and it is not even relieved by palliative chemotherapy. A double-blind, randomized clinical trial regarding the treatment of nonproductive cough was performed in 140 adults with primary lung cancer or metastatic cancer of the lungs. The therapeutic efficacy and the tolerability of a 7-day treatment with levodropropizine drops (75 mg t.i.d.) were evaluated in comparison with dihydrocodeine drops (10 mg t.i.d.; 7 days). Efficacy was assessed on the basis of cough severity scores, number of night awakenings due to cough, and overall estimate of antitussive efficacy. Tolerability was evaluated by laboratory results, vital signs and any adverse event occurring during the clinical trial, including presence or absence of somnolence. Subjective cough severity was significantly reduced during treatment with either levodropropizine and dihydrocodeine, the antitussive effect and its time-profile being similar for both drugs. Also, according to the investigator's evaluation, both levodropropizine and dihydrocodeine produced a significant decrease in cough severity. Concurrently with the relief of cough, the number of night awakenings was decreased significantly by both drugs, with no difference between the two treatments. No change in laboratory test values was considered clinically relevant, and vital signs were not clinically affected. The number of patients reporting adverse events was similar in the levodropropizine (n=6) and dihydrocodeine (n=4) group. However, the percentage of patients experiencing somnolence in the group receiving levodropropizine (8%) was significantly lower as compared with that of the dihydrocodeine group (22%). These results confirm the antitussive effectiveness of levodropropizine and suggest a more favourable benefit/risk profile when compared to dihydrocodeine.