Merck
Effect of selected triterpenoids on chronic dermal inflammation.
European journal of pharmacology (1997-11-05)
S Máñez, M C Recio, R M Giner, J L Ríos
ABSTRACT
The activity of four natural triterpenoids on a 12-O-tetradecanoylphorbol-13-acetate multiple-dose model of skin chronic inflammation was studied. Erythrodiol and ursolic acid were significantly effective. The most important features concerning structure-activity relationship and previous data on the effect of these triterpenoids on other inflammatory conditions are discussed.
Product Description
Supelco
Erythrodiol, analytical standard