Merck
Effects of levodropropizine on vagal afferent C-fibres in the cat.
British journal of pharmacology (1996-03-01)
H Shams, L Daffonchio, P Scheid
ABSTRACT
1. Levodropropizine (LVDP) is an effective antitussive drug. Its effects on single-unit discharge of vagal afferent C-fibres were tested in anaesthetized cats to assess whether an inhibition of vagal C-fibres is involved in its antitussive properties. Vagal C-fibres, identified by their response to phenylbiguanide (PBG), were recorded via suction electrodes from the distal part of the cut vagus. Based on their response to lung inflation, C-fibres were classified as pulmonary (19 fibres) or non-pulmonary (6 fibres). 2. PBG increased the discharge rate of both C-fibre types and activated a respiratory reflex causing apnoea. This reflex was abolished when the second vagus nerve was cut as well, while PBG-mediated stimulation of the C-fibres was not affected by vagotomy. 3. LVDP was administered intravenously and the C-fibre response to PBG was compared with that before administration of the drug. LVDP reduced both the duration of apnoea and the response of the C-fibre to PBG. 4. Comparison of the C-fibre responses to PBG and to a mixture of PBG and LVDP revealed that the period of apnoea was shortened and the discharge rate of the C-fibre reduced when LVDP was present. 5. The LVDP-induced inhibition of the C-fibre response to PBG was on average 50% in pulmonary and 25% in non-pulmonary fibres. 6. These results suggest that LVDP significantly reduces the response of vagal C-fibres to chemical stimuli. It is, thus, likely that the antitussive effect of LVDP is mediated through its inhibitory action on C-fibres.