Merck
  • Protective effects of the lazaroid U74500A and lidoflazine on liver preservation with UW solution.
Protective effects of the lazaroid U74500A and lidoflazine on liver preservation with UW solution.
Transplant international : official journal of the European Society for Organ Transplantation (1993-01-01)
J Jacobsson, R Sundberg, H L Rilo, A Gasbarrini, T E Starzl, D Van Thiel
ABSTRACT
The effect of adding a 21-aminosteroid, U74500A, and a Ca2+ antagonist, lidoflazine, alone and together to UW solution was assessed in a rat liver preservation model. Following preservation, the livers were reperfused using a closed circuit, and the release of hepatocellular enzymes (ASAT, ALAT, and LDH) into the perfusate was determined with increasing time. Both drugs reduced the amount of enzymes lost from the liver. The combination of the two drugs was better than either drug alone. These data suggest that both agents may be of value in organ preservation for clinical liver transplantation.
Sigma-Aldrich
Lidoflazine, ≥98% (HPLC), powder