- A comparative study on cytotoxicities and biochemical properties of anthraquinone mycotoxins emodin and skyrin from Penicillium islandicum Sopp.
A comparative study on cytotoxicities and biochemical properties of anthraquinone mycotoxins emodin and skyrin from Penicillium islandicum Sopp.
Toxicology letters (1984-02-01)
K Kawai, T Kato, H Mori, J Kitamura, Y Nozawa
PMID6320499
ABSTRACT
The anthraquinone mycotoxins emodin and skyrin were examined for the inhibitory effects on murine leukemia L1210 culture cells, oxidative phosphorylation of rat liver mitochondria, and Na+, K+-activated ATPase activity of rat brain microsomes to find the differences between their modes of toxic action. Skyrin exhibited a stronger inhibitory effect than emodin on the growth of L1210 culture cells. Emodin showed a stronger uncoupling effect than skyrin on mitochondrial respiration. Skyrin inhibited Na+, K+-activated ATPase activity of rat brain microsomes but emodin did not inhibit.