Merck
  • Amphiphilic N-(2,3-dihydroxypropyl)-chitosan-cholic acid micelles for paclitaxel delivery.
Amphiphilic N-(2,3-dihydroxypropyl)-chitosan-cholic acid micelles for paclitaxel delivery.
Carbohydrate polymers (2013-04-03)
Zheng Pan, Yunling Gao, Linseng Heng, Yi Liu, Gan Yao, Yun Wang, Yuping Liu
ABSTRACT
Self-assembled amphiphilic N-(2,3-dihydroxypropyl)-chitosan-cholic acid (DHP-CS-CHO) micelle was prepared as a carrier for paclitaxel. DHP-CS-CHO was synthesized by grafted small molecules cholic acid and glycidol onto primary amine group of chitosan, respectively. The DHP-CS-CHO formed uniform micelles (size=212.4±3.1 nm) with a low critical micelle concentration (0.024 mg/ml) in PBS. Hydrophobic anticancer drug, paclitaxel (PTX), was easily encapsulated into chitosan derivative micelles by a dialysis method with loading efficiency up to 80%. The PTX loaded micelles released the drug in a sustained manner more than a week in PBS containing 0.1% (w/v) Tween 80 at 37°C. In vitro antitumor experiment demonstrated that PTX loaded chitosan derivative micelles could inhibit MCF-7 cell growth and induce its apoptosis. These results suggested that DHP-CS-CHO may be a promising carrier for the anticancer drug PTX.
Sigma-Aldrich
Cholic acid, Vetec, reagent grade, 98%
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%