Merck
  • The low doses effect of experimental zearalenone (ZEN) intoxication on the presence of Ca2+ in selected ovarian cells from pre-pubertal bitches.
The low doses effect of experimental zearalenone (ZEN) intoxication on the presence of Ca2+ in selected ovarian cells from pre-pubertal bitches.
Polish journal of veterinary sciences (2013-02-09)
M Gajecka, B Przybylska-Gornowicz
ABSTRACT
The objective of this study was to determine the effect of 42-day ZEN intoxication on the presence of Ca2+ in selected ovarian cells from beagle bitches, using the potassium pyroantimonate (PPA) method. Samples were collected from 30 clinically healthy, pre-pubertal, genetically homogeneous animals. The bitches were divided into three groups of 10 animals each: experimental group I--50 microg ZEN/kg BW (100% NOAEL) administered once daily per os; experimental group II--75 microg ZEN/kg BW (150% NOAEL) administered once dailyper os; control group--placebo containing no ZEN administered per os. An electron microscopic analysis revealed that cells died due to apoptosis, depending on the ZEN dose and the type of cells exposed to intoxication. Lower ZEN doses led to apoptosis-like changes in the cells. Cell death was a consequence of excess Ca2+ accumulation in the mitochondria, followed by cell dysfunction and a decrease in or the absence of mitochondrial metabolic activity in oocytes, follicle cells and interstitial cells in experimental bitches.
Product Description
Supelco
Zearalenone solution, 100 μg/mL in acetonitrile, analytical standard