Merck
TAK1 is required for dermal wound healing and homeostasis.
The Journal of investigative dermatology (2013-01-24)
Fen Guo, James Hutchenreuther, David E Carter, Andrew Leask
ABSTRACT
Dermal connective tissue is a supportive structure required for skin's barrier function; dysregulated dermal homeostasis results in chronic wounds and fibrotic diseases. The multifunctional cytokine transforming growth factor (TGF) β promotes connective tissue deposition, repair, and fibrosis. TGF-β acts through well-defined canonical pathways; however, the non-canonical pathways through which TGF-β selectively promotes connective tissue deposition are unclear. In dermal fibroblasts, we show that inhibition of the non-canonical TGF-β-activated kinase 1 (TAK1) selectively reduced the ability of TGF-β to induce expression of a cohort of wound healing genes, such as collagens, CCN2, TGF-β1, and IL-6. Fibroblast-specific TAK1-knockout mice showed impaired cutaneous tissue repair and decreased collagen deposition, α-smooth muscle actin and CCN2 expression, proliferating cell nuclear antigen staining, and c-Jun N-terminal kinase and p38, but not Smad3, phosphorylation. TAK1-deficient fibroblasts showed reduced cell proliferation, migration, cell attachment/spreading, and contraction of a floating collagen gel matrix. TAK1-deficient mice also showed progressively reduced skin thickness and collagen deposition. Thus, TAK1 is essential for connective tissue deposition in the dermis.
Product Description
Supelco
Zearalenone solution, 100 μg/mL in acetonitrile, analytical standard