Merck
Template-free pseudomorphic synthesis of tungsten carbide nanorods.
Small (Weinheim an der Bergstrasse, Germany) (2012-07-26)
Ya Yan, Lan Zhang, Xiaoying Qi, Hao Song, Jing-Yuan Wang, Hua Zhang, Xin Wang
ABSTRACT
A unique nanorod-structured tungsten carbide material with high specific surface area of 198 m(2) g(-1) is successfully synthesized for the first time by pseudomorphic transformation of chemically synthesized WO(3) nanorods through a high-temperature method. An electrocatalyst composed of Pt nanoparticles supported on WC nanorods demonstrates higher electrocatalytic activity for methanol electro-oxidation, better tolerance to CO poisoning, and superior performance for cathodic electrocatalytic hydrogen evolution than a Pt/C catalyst. This work provides a novel method to synthesize high-surface-area nanorod-structured WC materials by preparing oxide precursors with the desired external morphology, thus offering great potential for a broad range of applications of these materials in related reaction systems.
Sigma-Aldrich
Tungsten(IV) carbide, powder, 2 μm, ≥99%