Merck
Neuroimaging: a scanner, colourfully.
Current biology : CB (2012-04-14)
Jonathan P Roiser, Geraint Rees
ABSTRACT
Two recent studies report changes in human brain responses after exposure to psilocybin, the active ingredient of hallucinogenic mushrooms. Psilocybin increased sensory cortex responses during emotional recollection, but decreased resting-state blood flow in prefrontal cortex, with potential implications for treating depression.