Merck
  • Surfactant properties of ionic liquids containing short alkyl chain imidazolium cations and ibuprofenate anions.
Surfactant properties of ionic liquids containing short alkyl chain imidazolium cations and ibuprofenate anions.
Physical chemistry chemical physics : PCCP (2011-07-30)
Corine Tourné-Péteilh, Jean-Marie Devoisselle, André Vioux, Patrick Judeinstein, Martin In, Lydie Viau
ABSTRACT
Interfacial tension, electrical conductivity, NMR self-diffusion and DLS experiments have been used to investigate the self-aggregation in water of ionic liquids associating an ibuprofenate anion and 1-alkyl-3-methylimidazolium [C(n)MIm](+) (n = 4, 6, 8) cations. Despite the short alkyl chain on imidazolium cations (n ≤ 8), these ionic liquids exhibit particularly low Critical Aggregation Concentrations (CAC), significantly lower than their parent 1-alkyl-3-methylimidazolium chloride salts. This behaviour is attributed to the formation of catanionic pairs between ibuprofenate and imidazolium.