Merck
  • Direct effects of fascaplysin on human umbilical vein endothelial cells attributing the anti-angiogenesis activity.
Direct effects of fascaplysin on human umbilical vein endothelial cells attributing the anti-angiogenesis activity.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2009-11-26)
Y L Zheng, X L Lu, J Lin, H M Chen, X J Yan, F Wang, W F Xu
ABSTRACT
Novel anti-angiogenesis activity of fascaplysin via VEGF blockage was recently revealed by our previous study in addition to the reported cyclin-dependent kinase 4 (CDK4) selective inhibition. To uncover more details of this pharmacologically prospective property, this study further investigated whether fascaplysin had direct anti-proliferation effects on human umbilical vein endothelial cells (HUVEC), which might be contributing to anti-angiogenesis. The results showed that G1 cell cycle arrest was induced by 2.6 μM fascaplysin in a time-dependent manner, and exhibited more sensitive than hepatocarcinoma cells BeL-7402 and Hela cells. Approximately 56.09 ± 2.63% of the cells were arrested at the G1 phase after 24h, and 64.94 ± 2.07% after 36 h, comparing to the 22.82 ± 1.2% in methanol treated cells. Apoptosis of HUVEC cells was induced by 1.3 μM fascaplysin and indicated by the sub-G1, Hoechst staining, terminal deoxynucleotidyl transferase dUTP-mediated nicked end labeling (TUNEL) assay, and annexin-V and propidium (PI) label. This apoptosis response was further confirmed by the detection of active caspase-3 and by western blotting using antibodies against Bax, Bcl-2, procaspase-8, and Bid, indicating that apoptosis in HUVEC cells may involve a mitochondria pathway, by the demonstration of an increase in the Bax/Bcl-2 ratio. Together, our results suggest that the anti-angiogenesis activity of fascaplysin is through the direct effects of cell cycle arrest and apoptosis on HUVEC.