Merck
Ethidium bromide: destruction and decontamination of solutions.
Analytical biochemistry (1987-05-01)
G Lunn, E B Sansone
ABSTRACT
Ethidium bromide in water, TBE buffer, Mops buffer, and cesium chloride solution may be completely degraded by reaction with sodium nitrite and hypophosphorous acid. Only non-mutagenic reaction mixtures were produced. Destruction was greater than 99.8% in all cases; the limit of detection was 0.5 micrograms ethidium bromide per milliliter of solution. Ethidium bromide also may be removed completely from the above solutions by using Amberlite XAD-16 resin. The limit of detection was 0.05 micrograms ethidium bromide per milliliter of solution (0.27 micrograms/ml when cesium chloride solution was used).