Merck
  • Reconstitution of ThiC in thiamine pyrimidine biosynthesis expands the radical SAM superfamily.
Reconstitution of ThiC in thiamine pyrimidine biosynthesis expands the radical SAM superfamily.
Nature chemical biology (2008-10-28)
Abhishek Chatterjee, Yue Li, Yang Zhang, Tyler L Grove, Michael Lee, Carsten Krebs, Squire J Booker, Tadhg P Begley, Steven E Ealick
ABSTRACT
4-Amino-5-hydroxymethyl-2-methylpyrimidine phosphate (HMP-P) synthase catalyzes a complex rearrangement of 5-aminoimidazole ribonucleotide (AIR) to form HMP-P, the pyrimidine moiety of thiamine phosphate. We determined the three-dimensional structures of HMP-P synthase and its complexes with the product HMP-P and a substrate analog imidazole ribotide. The structure of HMP-P synthase reveals a homodimer in which each protomer comprises three domains: an N-terminal domain with a novel fold, a central (betaalpha)(8) barrel and a disordered C-terminal domain that contains a conserved CX(2)CX(4)C motif, which is suggestive of a [4Fe-4S] cluster. Biochemical studies have confirmed that HMP-P synthase is iron sulfur cluster-dependent, that it is a new member of the radical SAM superfamily and that HMP-P and 5'-deoxyadenosine are products of the reaction. Mössbauer and EPR spectroscopy confirm the presence of one [4Fe-4S] cluster. Structural comparisons reveal that HMP-P synthase is homologous to a group of adenosylcobalamin radical enzymes. This similarity supports an evolutionary relationship between these two superfamilies.