- Fragrance material review on alpha-iso-methylionone.
Fragrance material review on alpha-iso-methylionone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-27)
A Lapczynski, J Lalko, V T Politano, D McGinty, S Bhatia, C S Letizia, A M Api
PMID18037210
ABSTRACT
A toxicologic and dermatologic review of alpha-iso-methylionone when used as a fragrance ingredient is presented.
MATERIALS