Merck
  • Effect of drug loading and molecular weight of cellulose acetate propionate on the release characteristics of theophylline microspheres.
Effect of drug loading and molecular weight of cellulose acetate propionate on the release characteristics of theophylline microspheres.
Pharmaceutical research (1991-11-01)
A J Shukla, J C Price
ABSTRACT
Microspheres with 40, 50, and 60% drug loading of anhydrous theophylline core material were prepared by the emulsion-solvent evaporation method. Three different molecular weights of cellulose acetate propionate were used as encapsulating polymers. The geometric mean diameter of the microspheres increased with drug loading for all polymers. Dissolution rate for a given particle size fraction also increased with drug loading for all polymers. Higuchi/Baker-Lonsdale spherical matrix dissolution kinetics were followed by narrow particle size fractions of the microspheres. A linear relationship between the T-50% (time required for 50% of the drug to be released) and the square of microsphere diameter was observed with all three molecular weights of the encapsulants. The slowest drug release was obtained with the high molecular weight polymer, which also produced the smoothest microspheres.
Sigma-Aldrich
Cellulose acetate propionate, average Mn ~15,000 by GPC, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Cellulose acetate propionate, average Mn ~75,000 by GPC, powder
Pricing and availability is not currently available.