Merck
  • Hyperammonemia acts synergistically with lipopolysaccharide in inducing changes in cerebral hemodynamics in rats anaesthetised with pentobarbital.
Hyperammonemia acts synergistically with lipopolysaccharide in inducing changes in cerebral hemodynamics in rats anaesthetised with pentobarbital.
Journal of hepatology (2007-05-29)
Hans R Pedersen, Helmer Ring-Larsen, Niels Vidiendal Olsen, Fin Stolze Larsen
ABSTRACT
The aim was to determine the effect of ammonia (NH(3)) and lipopolysaccharide (LPS) alone or in combination, on cerebral blood flow (CBF) and intracranial pressure (ICP) in the rat. Since amiloride-sensitive-ion-pathways in the blood-brain barrier (BBB) modulate CBF, we also aimed to test if Na(+)/H(+)-inhibitors could prevent this possible synergism between NH(3) and LPS. In experiment A, four groups of rats received ammonium acetate (140 micromol/kg/min) or saline, each of them associated with either vehicle or LPS (2 mg/kg). In experiments B and C, rats received similar treatments after having received amiloride (30 mg/kg) or 5-(N-methyl-N-isobutyl)-amiloride (MIA, 5 mg/kg). Plasma tumor-necrosis-factor-alpha (TNF-alpha), ICP (via a cisterna magna catheter) and CBF (by laser-Doppler flowmetry) were measured. An increase in ICP and CBF within 60 min was observed only in rats that received NH(3) together with LPS as compared to any other group (P<0.01), which could be prevented by amiloride (P<0.05), but not by MIA. Both amiloride and MIA decreased the plasma TNF-alpha concentration. In rats anaesthetised with pentobarbital NH(3) infusion aggravates a LPS induced rise in ICP and induces an increase in CBF less clearly seen with LPS alone. This effect is prevented by the non-specific Na(+)/H(+) inhibitor amiloride, but not by MIA, a specific inhibitor of Na(+)/H(+) exchanger. Thus, the synergistic effect of NH(3) and LPS seems mediated by other amiloride-sensitive-ion-pathways in the BBB than the Na(+)/H(+) exchanger.