Merck
  • Preparation and antibacterial activity of copper and cobalt complexes of 4-chloro-3-nitrobenzoate with a nitrogen donor ligand.
Preparation and antibacterial activity of copper and cobalt complexes of 4-chloro-3-nitrobenzoate with a nitrogen donor ligand.
Chemical & pharmaceutical bulletin (2007-03-03)
Ahmad Toufic Kabbani, Hassan Hasan Hammud, Amer Mohammed Ghannoum
ABSTRACT
Copper and cobalt complexes with 4-chloro-3-nitrobenzoate (ClNBz) and the nitrogen ligands 1,3-diaminopropane (1,3-DAP) or o-phenylenediamine (o-PDA), were prepared and characterized. The complexes [Cu(ClNBz)2(1,3-DAP)] (1), [Cu(ClNBz)(o-PDA)]Cl (2), [Co(ClNBz)2(1,3-DAP)] (3) and [Co(ClNBz)2(o-PDA)2] (4) were characterized by FTIR, UV-Visible absorption, elemental analysis and thermal analysis. Complex [Cu(ClNBz)(o-PDA)]Cl (2) shows high antibacterial activity as indicated by its ability to inhibit the growth of Staphylococcus aureus, Enterococcus faecalis.
Sigma-Aldrich
4-Chloro-3-nitrobenzoic acid, 98%