Merck
  • C-Npys (S-3-nitro-2-pyridinesulfenyl) and peptide derivatives can inhibit a serine-thiol proteinase activity from Paracoccidioides brasiliensis.
C-Npys (S-3-nitro-2-pyridinesulfenyl) and peptide derivatives can inhibit a serine-thiol proteinase activity from Paracoccidioides brasiliensis.
Biochemical and biophysical research communications (2007-03-03)
Alisson L Matsuo, Adriana K Carmona, Luiz S Silva, Carlos E L Cunha, Ernesto S Nakayasu, Igor C Almeida, Maria A Juliano, Rosana Puccia
ABSTRACT
The inhibitory capacity of C-Npys (S-[3-nitro-2-pyridinesulfenyl]) derivatives over thiol-containing serine proteases has never been tested. In the present work we used an extracellular serine-thiol proteinase activity from the fungal pathogen Paracoccidioides brasiliensis (PbST) to describe a potent inhibitory capacity of Bzl-C(Npys)KRLTL-NH(2) and Bzl-MKRLTLC(Npys)-NH(2). The assays were performed with PbST enriched upon affinity chromatography in a p-aminobenzamidine (pABA)-Sepharose column. Although PbST can cleave the fluorescence resonance energy transfer peptide Abz-MKRLTL-EDDnp between L-T, the C(Npys) derivatives were not substrates nor were they toxic in a cell detachment assay, allowing therapeutic use. The best inhibitor was Bzl-C(Npys)KRLTL-NH(2) (K(i)=16nM), suggesting that the peptide sequence promoted a favorable interaction, especially when C(Npys) was placed at a further position from the L-T bond, at the N-terminus. Inhibition was completely reverted with dithioerythritol, indicating that it was due to the reactivity of the C(Npys) moiety with a free SH- group.
Sigma-Aldrich
3-Nitro-2-pyridinesulfenyl chloride, 95%
Pricing and availability is not currently available.