Merck
  • Effects of some benzoxazinoids on in vitro growth of Cephalosporium gramineum and other fungi pathogenic to cereals and on Cephalosporium stripe of winter wheat.
Effects of some benzoxazinoids on in vitro growth of Cephalosporium gramineum and other fungi pathogenic to cereals and on Cephalosporium stripe of winter wheat.
Journal of agricultural and food chemistry (2006-02-16)
Stefan Martyniuk, Anna Stochmal, Francisco A Macías, David Marín, Wieslaw Oleszek
ABSTRACT
The benzoxazolinones benzoxazolin-2(3H)-one (BOA) and 6-methoxybenzoxazolin-2(3H)-one (MBOA) and selected degradation products of these compounds were examined for their in vitro antifungal activity against Cephalosporium gramineum, Gaeumannomyces graminis var. graminis, and Fusarium culmorum. BOA was also applied to the soil-incorporated inoculum of C. gramineum to test its capability of reducing Cephalosporium stripe disease in winter wheat. MBOA reduced the mycelial growth of G. graminis var. tritici, C. gramineum, and F. culmorum by 50% (EC50) at the concentrations of 77, 134, and 271 microg/mL of corn meal agar, respectively, and the corresponding BOA EC50 values for the fungi were 11, 189, and 456 microg/mL. BOA degradation products 2-amino-3H-phenoxazin-3-one (APO), 2-acetylamino-3H-phenoxazin-3-one (AAPO), and o-aminophenol (o-AP) were much more inhibitory to the growth of C. gramineum and G. graminis var. tritici than the parent compounds. APO, AAPO, and o-AP EC50 values were found to be as low as 0.58, 4.57, and 1.4 microg/mL, respectively, for C. gramineum and 0.78, 2.18, and 0.80 microg/mL for G. graminis var. tritici. These compounds applied at the corresponding concentrations did not significantly affect the mycelial growth of F. culmorum. The treatment of C. gramineum inoculum with a 1% water solution of BOA resulted in a significant reduction infection of winter wheat with C. gramineum as compared to the control with the untreated inoculum,but this treatment was not as effective as the application of a commercial fungicide.
Sigma-Aldrich
6-Methoxy-2-benzoxazolinone, 97%