- Gender differences in cocaine pharmacokinetics in CF-1 mice.
Gender differences in cocaine pharmacokinetics in CF-1 mice.
Hepatocellular damage is thought to occur as a result of cytochrome P450-mediated oxidation of cocaine to norcocaine (NC), a precursor of the hepatotoxic nitrosonium ion. However, this damage occurs only in male mice, with females exhibiting minimal biochemical and histological signs of hepatocellular stress. The objective of this study was to determine the plasma time course and tissue disposition of cocaine and its metabolites to further investigate the role that metabolism may play in the gender difference observed. Male and female CF-1 mice were orally administered 20mg/kg cocaine hydrochloride once daily for 7 days. Blood samples were withdrawn at various time points post-injection and analyzed for cocaine and its metabolites benzoylecgonine (BE), norcocaine, ecgonine methyl ester (EME), and ecgonine (E). In addition, tissue concentrations of cocaine and its metabolites were determined in liver, heart, brain, and kidney tissue. The results demonstrated that the plasma elimination half-life of cocaine is nearly three times longer in males versus females. Non-hepatotoxic hydrolysis metabolites BE, EME, and E were higher in female tissues while norcocaine was detected in tissues of male animals only. This study revealed that differences in cocaine pharmacokinetics and the resultant differences in the biodisposition of cocaine and its metabolites in tissues contribute to the mechanism of gender difference seen in cocaine hepatotoxicity.