Merck
  • Studies on the human metabolism and the toxicologic detection of the cough suppressant dropropizine in urine using gas chromatography-mass spectrometry.
Studies on the human metabolism and the toxicologic detection of the cough suppressant dropropizine in urine using gas chromatography-mass spectrometry.
Therapeutic drug monitoring (2004-07-17)
Roland F Staack, Denis S Theobald, Hans H Maurer
ABSTRACT
Studies are described on the metabolism and the toxicologic analysis of the nonopioid cough suppressant dropropizine [R,S-3-(4-phenyl-1-piperazinyl)1,2-propandiol, DRO] in human urine using gas chromatography-mass spectrometry (GC-MS). The metabolism studies showed that DRO was metabolized in humans mainly by hydroxylation of the aromatic ring, by N-dealkylation of the parent drug and of the hydroxyl-metabolite to the corresponding N-phenylpiperazines, and by degradation of the piperazine moiety. The authors' systematic toxicologic analysis (STA) procedure using full-scan GC-MS after acid hydrolysis, liquid-liquid extraction, and microwave-assisted acetylation allowed the unambiguous detection of DRO and its above-mentioned metabolites in human urine up to about 32 hours after intake of a single common therapeutic dose. The target analytes were found to be the parent compound DRO (earlier phase of excretion) and the hydroxylated metabolite para-hydroxy-DRO (later phase of excretion). Both allowed unambiguous detection of an intake of DRO and also differentiation from other phenylpiperazine derivatives.