Merck
  • Specific nonpeptide inhibitors of puromycin-sensitive aminopeptidase with a 2,4(1H,3H)-quinazolinedione skeleton.
Specific nonpeptide inhibitors of puromycin-sensitive aminopeptidase with a 2,4(1H,3H)-quinazolinedione skeleton.
Chemical & pharmaceutical bulletin (2003-11-06)
Hiroki Kakuta, Aya Tanatani, Kazuo Nagasawa, Yuichi Hashimoto
ABSTRACT
Potent, specific, chemically stable and non-peptide/small-molecular inhibitors of puromycin-sensitive aminopeptidase, such as 3-(2,6-diethylphenyl)-2,4(1H,3H)-quinazolinedione (PAQ-22, 5), were prepared by the structural development of a potent PSA inhibitor, 2-(2,6-diethylphenyl)-1,2,3,4-tetrahydroisoquinoline-1,3-dione (PIQ-22, 4). The design was carried out partly by applying electrostatic potential field information obtained from PIQ-22 (4) and its derivatives based on thalidomide (2). This information revealed that a positive electrostatic potential field around the benzylic methylene in the tetrahydroisoquinoline ring is necessary for potent activity. Lineweaver-Burk plot analysis showed that PAQ-22 (5) and its derivatives inhibit puromycin-sensitive aminopeptidase (PSA) in a non-competitive manner. These potent and specific PSA inhibitors showed dose-dependent cell invasion-inhibitory activity in a Matrigel assay using mouse melanoma B16F10/L5 cells, in spite of their low cell toxicity.
Sigma-Aldrich
Benzoyleneurea, 97%