Merck
  • Synthesis and biological activity of some 5-substituted aminomethyl-8-hydroxyquinoline-7-sulphonic acids.
Synthesis and biological activity of some 5-substituted aminomethyl-8-hydroxyquinoline-7-sulphonic acids.
Journal of chemical technology and biotechnology (Oxford, Oxfordshire : 1986) (1990-01-01)
A S Yanni, A M Mohharam
ABSTRACT
5-Aryl (or alkyl)-8-hydroxyquinoline-7-sulphonic acids have been prepared by the Mannich reaction of 8-hydroxyquinoline-7-sulphonic acid with primary and secondary amines. Their bactericidal activities have been determined.