- Design and synthesis of the tumor-activated prodrug of dihydropyrimidine dehydrogenase (DPD) inhibitor, RO0094889 for combination therapy with capecitabine.
Design and synthesis of the tumor-activated prodrug of dihydropyrimidine dehydrogenase (DPD) inhibitor, RO0094889 for combination therapy with capecitabine.
Bioorganic & medicinal chemistry letters (2003-03-06)
Kazuo Hattori, Yasunori Kohchi, Nobuhiro Oikawa, Hitomi Suda, Masako Ura, Tohru Ishikawa, Masanori Miwa, Mika Endoh, Hiroyuki Eda, Hiromi Tanimura, Akira Kawashima, Ikuo Horii, Hideo Ishitsuka, Nobuo Shimma
PMID12617910
ABSTRACT
A series of tumor-activated prodrugs of the inhibitors of dihydropyrimidine dehydrogenase (DPD), an enzyme catabolizing 5-fluorouracil (5-FU: 4g), has been designed and synthesized. RO0094889 (11c) is a prodrug of 5-vinyluracil (4c), a known DPD inhibitor, and was designed to generate 4c selectively in tumor tissues by sequential conversion of 11c by three enzymes: esterase, cytidine deaminase and thymidine phosphorylase, the latter two of which are known to be highly expressed in various tumor tissues. When capecitabine (1), a tumor-activated prodrug of 5-FU, was co-administered orally with 11c, 5-FU in tumor tissues was significantly increased with only a slight increase of 5-FU in plasma as compared with oral capecitabine alone.