Merck
  • Impurities in a morphine sulfate drug product identified as 5-(hydroxymethyl)-2-furfural, 10-hydroxymorphine and 10-oxomorphine.
Impurities in a morphine sulfate drug product identified as 5-(hydroxymethyl)-2-furfural, 10-hydroxymorphine and 10-oxomorphine.
Journal of pharmaceutical sciences (2003-02-15)
Seán S Kelly, Padraig M Glynn, Stuart J Madden, David H Grayson
ABSTRACT
Stability testing of morphine sulfate formulated with nonpareil sugar seeds (consisting of sucrose and starch) and fumaric acid revealed the formation of the three impurities 5-(hydroxymethyl)-2-furfural, 10-hydroxymorphine and 10-oxomorphine. 5-(Hydroxymethyl)-2-furfural was isolated via semipreparative HPLC utilizing volatile mobile phase constituents and was identified by analysis of its HRMS and NMR spectra. 10-Hydroxymorphine and 10-oxomorphine were obtained via semipreparative HPLC and subsequent removal of ion-pair reagents using an anion exchange resin, or by solid phase extraction, and identified by spectroscopic analysis followed by comparison with authentic materials. 5-(Hydroxymethyl)-2-furfural is a degradation product of hexose sugars, and its formation in the presence of morphine sulfate formulated with fumaric acid suggests that caution should be exercised when including nonpareil seeds in formulations that contain acidic drug salts and/or acid excipients. The preliminary results of tests on the interaction of acidic salts of some other drugs with nonpareil seeds are presented.