- Fatality caused by a combined trimipramine-citalopram intoxication.
Fatality caused by a combined trimipramine-citalopram intoxication.
Forensic science international (2000-02-09)
F Musshoff, P Schmidt, B Madea
PMID10664899
ABSTRACT
A 53-year-old woman who was diagnosed as suffering from depression was found dead in her bed. The autopsy revealed no morphological changes sufficient to explain death. Toxicological analysis was performed and the drugs trimipramine (2.33 mg/l), citalopram (4.81 mg/l) and zolpidem (0.07 mg/l) were identified in the femoral blood. A combined drug intoxication resulting in synergistic effects to cardiovascular disorders was proposed as the cause of death. An acute overdose and suicide was suggested by calculation of the parent drug to main metabolite ratios in femoral blood and liver tissue. The trimipramine to desmethyltrimipramine ratios were calculated to be 2.06 and 3.18, the citalopram to desmethylcitalopram ratios were 1.96 and 2.02.