Merck
  • O-(fluoresceinylmethyl)hydroxylamine (OFMHA): a reagent for the preparation of fluorescent O-(fluoresceinylmethyl)oxime (FMO)-steroid conjugates.
O-(fluoresceinylmethyl)hydroxylamine (OFMHA): a reagent for the preparation of fluorescent O-(fluoresceinylmethyl)oxime (FMO)-steroid conjugates.
Steroids (1999-07-10)
M Adamczyk, Y Y Chen, J Grote, P G Mattingly
ABSTRACT
The 5 and 6-isomers of O-(fluoresceinylmethyl)hydroxylamine reacted with a representative sample of oxo-steroids (6-oxoestradiol, estrone, norethindrone, cortisol, progesterone, and digitoxin-dialdehyde) to produce O-(fluoresceinylmethyl)oxime conjugates in a single step in 24-84% yield after preparative high performance liquid chromotography.