- A de novo mutation affecting human TrkB associated with severe obesity and developmental delay.
A de novo mutation affecting human TrkB associated with severe obesity and developmental delay.
Nature neuroscience (2004-10-21)
Giles S H Yeo, Chiao-Chien Connie Hung, Justin Rochford, Julia Keogh, Juliette Gray, Shoba Sivaramakrishnan, Stephen O'Rahilly, I Sadaf Farooqi
PMID15494731
ABSTRACT
An 8-year-old male with a complex developmental syndrome and severe obesity was heterozygous for a de novo missense mutation resulting in a Y722C substitution in the neurotrophin receptor TrkB. This mutation markedly impaired receptor autophosphorylation and signaling to MAP kinase. Mutation of NTRK2, which encodes TrkB, seems to result in a unique human syndrome of hyperphagic obesity. The associated impairment in memory, learning and nociception seen in the proband reflects the crucial role of TrkB in the human nervous system.
Sigma-Aldrich
TrkB/Fc Chimera human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder