Merck
  • [Acute myocardial infarction mimicking takotsubo cardiomyopathy in a patient with myocardial bridging].
[Acute myocardial infarction mimicking takotsubo cardiomyopathy in a patient with myocardial bridging].
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego (2013-01-01)
Waldemar Elikowski, Małgorzata Małek, Małgorzata Pyda, Mieczysław Dziarmaga, Dariusz Angerer, Dominika Montewska
ABSTRACT
The authors present a case of a 70-year-old woman in whom clinical picture of an acute coronary syndrome with ST segment elevation was suggestive of takotsubo cardiomyopathy (TC). Chest pain, extensive ECG changes and typical TC left ventricular contraction pattern were preceded by emotional and physical stress, while in coronary angiography no atherosclerotic lesions were found. There was however left anterior descending coronary artery myocardial bridging with total systolic compression. Following treatment with beta-blocker and diltiazem, spectacular left ventricle function improvement, with near total recovery after 6 months was observed. Magnetic resonance imaging detected the presence of subendocardial late gadolinium enhancement indicative of postmyocardial scar.
Product Description
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)