Merck
  • Effectiveness of rifaximin and fluoroquinolones in preventing travelers' diarrhea (TD): a systematic review and meta-analysis.
Effectiveness of rifaximin and fluoroquinolones in preventing travelers' diarrhea (TD): a systematic review and meta-analysis.
Systematic reviews (2012-08-30)
Sanjin Alajbegovic, John W Sanders, Deborah E Atherly, Mark S Riddle
ABSTRACT
Recent developments related to a safe and effective nonabsorbable antibiotic, rifaximin, and identification of postinfectious irritable bowel syndrome as a frequent sequela call for a need to reconsider the value of primary prevention of traveler's diarrhea (TD) with antibiotics. Randomized, placebo-controlled, double-blind studies evaluating the effectiveness and safety of rifaximin or a fluoroquinolone chemoprophylaxis against TD were pooled using a random effects model and assessed for heterogeneity. The nine studies (four rifaximin and five fluoroquinolone) included resulted in pooled relative risk estimates of 0.33 (95% CI = 0.24-0.45, I2 = 3.1%) and 0.12 (95% CI = 0.07-0.20, I2 =0.0%), respectively. Similar rates of treatment emergent adverse events were found between antibiotic and placebo groups. This meta-analysis supports the effectiveness of antibiotics in preventing TD. However, further studies that include prevention of secondary chronic health outcomes among travelers to different geographic regions, and a formal risk-benefit analysis for antibiotic chemoprophylaxis, are needed.
Product Description
Supelco
Rifaximin, VETRANAL®, analytical standard