MilliporeSigma
  • Synthesis, radiolabeling, and bio-imaging of high-generation polyester dendrimers.
Synthesis, radiolabeling, and bio-imaging of high-generation polyester dendrimers.
Journal of the American Chemical Society (2009-02-26)
Matthew C Parrott, S Rahima Benhabbour, Chantal Saab, Jennifer A Lemon, Shannon Parker, John F Valliant, Alex Adronov
ABSTRACT
A series of aliphatic polyester dendrons, generations 1 through 8, were prepared with a core p-toluenesulfonyl ethyl (TSe) ester as an easily removable protecting group that can be efficiently replaced with a variety of nucleophiles. Using amidation chemistry, a tridentate bis(pyridyl)amine ligand which is known to form stable complexes with both Tc(I) and Re(I) was introduced at the dendrimer core. Metalation of the core ligand with (99m)Tc was accomplished for generations 5 through 7, and resulted in regioselective radiolabeling of the dendrimers. The distribution of the radiolabeled dendrimers was evaluated in healthy adult Copenhagen rats using dynamic small-animal single photon emission computed tomography (SPECT). The labeled dendrimers were cleanly and rapidly eliminated from the bloodstream via the kidneys with negligible nonspecific binding to organs or tissues being observed. These data were corroborated by a quantitative biodistribution study on the generation 7 dendrimer following necropsy. The quantitative biodistribution results were in excellent agreement with the data obtained from the dynamic SPECT images.
Sigma-Aldrich
2,2′-Dipyridylamine, 98%