Merck
  • Crystallographic studies of Homo sapiens ribosomal decoding A site complexed with aminoglycosides.
Crystallographic studies of Homo sapiens ribosomal decoding A site complexed with aminoglycosides.
Nucleic acids symposium series (2004) (2006-12-08)
Jiro Kondo, Boris François, Alexandre Urzhumtsev, Eric Westhof
ABSTRACT
Aminoglycosides are highly effective antibacterial drugs that decrease translation accuracy by binding to the aminoacyl-tRNA decoding site (A site) of 16S ribosomal RNA. On the other hand, they are highly toxic to mammals through kidney and ear-associated illnesses by binding to ribosomal A sites. To understand the mechanism of toxicity of aminoglycosides to mammals at atomic level, crystallographic studies have been carried out with a number of Homo sapiens mitochondrial and cytoplasmic A sites complexed with aminoglycosides. Several X-ray diffraction data sets were successfully collected. Initial phases of mitochondrial A site with tobramycin and cytoplasmic A site with paromomycin were derived by the molecular replacement method. Refinements of atomic parameters are now under progress.