Merck
  • Amination of tyrosine in liver cytosol protein of male F344 rats treated with 2-nitropropane, 2-nitrobutane, 3-nitropentane, or acetoxime.
Amination of tyrosine in liver cytosol protein of male F344 rats treated with 2-nitropropane, 2-nitrobutane, 3-nitropentane, or acetoxime.
Chemical research in toxicology (1998-01-23)
R S Sodum, E S Fiala
ABSTRACT
Previously, the secondary nitroalkane 2-nitropropane, a strong hepatocarcinogen in rats, had been shown to induce the formation of 8-aminoguanine in both DNA and RNA of rat liver through a sulfotransferase-mediated pathway. This pathway was postulated to convert the carcinogen into an aminating species [Sodum, R. S., et al. (1994) Chem. Res. Toxicol. 7, 344-351]. To submit this postulate to further test, we examined liver proteins of rats treated with 2-nitropropane, other carcinogenic secondary nitroalkanes, or the related rat liver tumorigen acetoxime for the presence of 3-aminotyrosine, the expected product of tyrosine amination. Using ion-pair and/or cation-exchange high-performance liquid chromatography with electrochemical detection, we found that the liver cytosolic proteins of these animals contained 0.1-1.5 mol of 3-aminotyrosine/10(3) mol of tyrosine. Treatment with the noncarcinogenic primary nitroalkane 1-nitropropane or with other primary nitroalkanes did not produce an analogous increase in the aminated amino acid (level of detection estimated at approximately 0.01 mol/10(3) mol of tyrosine). To our knowledge, this is the first report of the modification of protein tyrosine in vivo by a carcinogen. In vitro studies with acetoxime-O-sulfonate and hydroxylamine-O-sulfonate showed that these proposed intermediates in the activation pathway of 2-nitropropane react with guanosine to give 8-aminoguanosine, N1-aminoguanosine, and 8-oxoguanosine and also react with tyrosine to give 3-aminotyrosine and 3-hydroxytyrosine. The in vitro amination and oxidation of guanosine at C8 were also produced by acetophenoxime-O-sulfonate and 2-heptanoxime-O-sulfonate. These results provide additional evidence for the production of a reactive species capable of aminating nucleic acids and proteins from 2-nitropropane and other carcinogenic secondary nitroalkanes by a pathway involving oxime- and hydroxylamine-O-sulfonates as intermediates.
Sigma-Aldrich
Acetone oxime, 98%
Sigma-Aldrich
Acetone oxime, purum, ≥98.0% (GC)
Sigma-Aldrich
Acetone oxime, Vetec, reagent grade, 98%