Merck
  • Induction of manganese superoxide dismutase by thyroid stimulating hormone in rat thyroid cells.
Induction of manganese superoxide dismutase by thyroid stimulating hormone in rat thyroid cells.
FEBS letters (1997-11-22)
S Nishida, T Nakano, S Kimoto, T Kusunoki, K Suzuki, N Taniguchi, K Murata, T T Tomura
ABSTRACT
Alterations in the superoxide dismutase (SOD) content of thyroid tissues occurring in association with thyroid dysfunction have been reported. In this study, the Mn-SOD content was found to increase in thyroid tissues of rats administered thyroid stimulating hormone (TSH) and in thyrocytes cultured in medium supplemented with TSH. Furthermore, in the thyroid glands of rats whose serum TSH level was elevated by inhibiting the synthesis of T3 and T4 by 6-methyl-2-thiouracil, the Mn-SOD increased as the TSH concentration increased. In the cultured thyrocytes, the increase in Mn-SOD induced by TSH was inhibited by the C-kinase inhibitor H7. These findings suggest the induction of Mn-SOD by TSH in thyroid cells and point to a role of C-kinase in this process, thereby indicating that a close relationship exists between the serum TSH level and the change in Mn-SOD content in thyrocytes with thyroid dysfunction.
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)