Merck
  • Controlled release of theophylline monohydrate from amylodextrin tablets: in vitro observations.
Controlled release of theophylline monohydrate from amylodextrin tablets: in vitro observations.
Pharmaceutical research (1994-04-01)
J Van der Veen, G H Te Wierik, L Van der Wal, A C Eissens, C F Lerk
ABSTRACT
Amylodextrin is a linear dextrin and can be produced by enzymatic hydrolysis of the alpha-1,6 glycosidic bonds of amylopectin. Tablets compacted from pure amylodextrin showed good binding properties and did not disintegrate in aqueous media. Extended and decreasing drug release rates were found for tablets of 300 mg with a diameter of 9 mm containing 70% amylodextrin and 30% theophylline monohydrate, when compacted at 5 kN. Almost-constant drug release rates were obtained for these tablets when compacted at 10 or 15 kN. Nearly constant drug release rates were also shown for amylodextrin tablets with a drug load up to 75% compacted at 10 kN. Both release rate and release profile could be adjusted by selecting tablet thickness and incorporation of either lactose as a highly soluble excipient or talc as a hydrophobic excipient.
Sigma-Aldrich
Starch, soluble, ACS reagent
Sigma-Aldrich
Starch, soluble, Vetec, reagent grade