Merck
Aminoacyl-RNA synthesis catalyzed by an RNA.
Science (New York, N.Y.) (1995-02-03)
M Illangasekare, G Sanchez, T Nickles, M Yarus
ABSTRACT
An RNA has been selected that rapidly aminoacylates its 2'(3') terminus when provided with phenylalanyl-adenosine monophosphate. That is, the RNA accelerates the same aminoacyl group transfer catalyzed by protein aminoacyl-transfer RNA synthetases. The best characterized RNA reaction requires both Mg2+ and Ca2+. These results confirm a necessary prediction of the RNA world hypothesis and represent efficient RNA reaction (> or = 10(5) times accelerated) at a carbonyl carbon, exemplifying a little explored type of RNA catalysis.
Sigma-Aldrich
2-Naphthoxyacetic acid, BioReagent, suitable for plant cell culture, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
2-Naphthoxyacetic acid, 98%
Pricing and availability is not currently available.