Merck
  • Blocking by 2,4,6-triaminopyrimidine of increased tight junction permeability induced by acetylcholine in the pancreas.
Blocking by 2,4,6-triaminopyrimidine of increased tight junction permeability induced by acetylcholine in the pancreas.
Biochimica et biophysica acta (1980-05-08)
J W Jansen, A M Fleuren-Jakobs, J J De Pont, S L Bonting
ABSTRACT
1. The permeability of the paracellular pathway in the isolated rabbit pancreas has been studied with the aid of 2,4,6-triaminopyrimidine. 2. Addition of 2,4,6-triaminopyrimidine (1--10 mM) to the bathing medium has no effect on the rate of fluid secretion or on protein, Na+, K+, Ca2+ and sucrose concentrations in the secreted fluid. 3. When 1 x 10(-5) M carbachol is also added to the 2,4,6-triaminopyrimidine-containing bathing medium, there is a marked reduction in the increase of the paracellular permeability for sucrose and Ca2+ found upon addition of carbachol alone. The enzyme secretion, induced by carbachol, is not affected. 4. The minimal concentration of 2,4,6-triaminopyrimidine in the bathing medium required to reach its maximal effect on the paracellular permeability is approx. 0.55 mM at pH 7.4. 5. The effect of 2,4,6-triaminopyrimidine on the paracellular permeability after carbachol stimulation is also present when 2,4,6-triaminopyrimidine is added 5 min after the addition of 1 x 10(-5) M carbachol. 6. 2,4,6-Triaminopyrimidine has no effect on the increases in enzyme secretion and sucrose permeability caused by 1 x 10(-8) pancreozymin C octapeptide. 7. 2,4,6-Triaminopyrimidine appears in the secreted fluid at a concentration of 50% of that in the bathing medium. Upon addition of 1 x 10(5) M carbachol this concentration increases up to 80%. 8. These results indicate that: (a) the increased paracellular permeability upon stimulation with carbachol is not caused by the enzyme secretion as such and (b) addition of 2,4,6-triaminopyrimidine prevents the carbachol-induced increase in permeability of a channel in the tight junction complex.
Sigma-Aldrich
2,4,6-Triaminopyrimidine, 97%
Pricing and availability is not currently available.