Merck
  • The reduction of hydroxamic acids with titanium(III) chloride: a tool for the characterization of siderophores.
The reduction of hydroxamic acids with titanium(III) chloride: a tool for the characterization of siderophores.
Analytical biochemistry (1983-08-01)
D J Kushner, T A Landry, M C Tyrrell, H A Akers
ABSTRACT
Hydroxamic acid siderophores were observed to be inactivated by exposure to titanium(III) chloride. To study the reaction, a series of eight model hydroxamic acids were prepared and reacted with titanium(III) chloride. The products were shown by ir and NMR comparisons with authentic compounds to be the corresponding amides. The reduction was found to require 2 mol of titanium(III) per mol of hydroxamic acid.
Sigma-Aldrich
Titanium(III) chloride, ReagentPlus®, powder
Sigma-Aldrich
Titanium(III) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(III) chloride solution, 10-15% TiCl3 basis