Merck
  • Nonsurgical management of heavy menstrual bleeding: a systematic review.
Nonsurgical management of heavy menstrual bleeding: a systematic review.
Obstetrics and gynecology (2013-05-03)
Kristen A Matteson, David D Rahn, Thomas L Wheeler, Elizabeth Casiano, Nazema Y Siddiqui, Heidi S Harvie, Mamta M Mamik, Ethan M Balk, Vivian W Sung
ABSTRACT
To compare the effectiveness of nonsurgical abnormal uterine bleeding treatments for bleeding control, quality of life (QOL), pain, sexual health, patient satisfaction, additional treatments needed, and adverse events. MEDLINE, Cochrane databases, and Clinicaltrials.gov were searched from inception to May 2012. We included randomized controlled trials of nonsurgical treatments for abnormal uterine bleeding presumed secondary to endometrial dysfunction and abnormal uterine bleeding presumed secondary to ovulatory dysfunction. Interventions included the levonorgestrel intrauterine system, combined oral contraceptive pills (OCPs), progestins, nonsteroidal anti-inflammatory drugs (NSAIDs), and antifibrinolytics. Gonadotropin-releasing hormone agonists, danazol, and placebo were allowed as comparators. Two reviewers independently screened 5,848 citations and extracted eligible trials. Studies were assessed for quality and strength of evidence. Twenty-six articles met inclusion criteria. For reduction of menstrual bleeding in women with abnormal uterine bleeding presumed secondary to endometrial dysfunction, the levonorgestrel intrauterine system (71-95% reduction), combined OCPs (35-69% reduction), extended cycle oral progestins (87% reduction), tranexamic acid (26-54% reduction), and NSAIDs (10-52% reduction) were all effective treatments. The levonorgestrel intrauterine system, combined OCPs, and antifibrinolytics were all superior to luteal-phase progestins (20% increase in bleeding to 67% reduction). The levonorgestrel intrauterine system was superior to combined OCPs and NSAIDs. Antifibrinolytics were superior to NSAIDs for menstrual bleeding reduction. Data were limited on other important outcomes such as QOL for women with abnormal uterine bleeding presumed secondary to endometrial dysfunction and for all outcomes for women with abnormal uterine bleeding presumed secondary to ovulatory dysfunction. For the reduction in mean blood loss in women with heavy menstrual bleeding presumed secondary to abnormal uterine bleeding presumed secondary to endometrial dysfunction, we recommend the use of the levonorgestrel intrauterine system over OCPs, luteal-phase progestins, and NSAIDs. For other outcomes (QOL, pain, sexual health, patient satisfaction, additional treatments needed, and adverse events) and for treatment of abnormal uterine bleeding presumed secondary to ovulatory dysfunction, we were unable to make recommendations based on the limited available data.
Sigma-Aldrich
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%