Merck
  • Analysis of plasticizers and synthetic musks in cosmetic and personal care products by matrix solid-phase dispersion gas chromatography-mass spectrometry.
Analysis of plasticizers and synthetic musks in cosmetic and personal care products by matrix solid-phase dispersion gas chromatography-mass spectrometry.
Journal of chromatography. A (2013-04-30)
Maria Llompart, Maria Celeiro, J Pablo Lamas, Lucia Sanchez-Prado, Marta Lores, Carmen Garcia-Jares
ABSTRACT
Matrix solid-phase dispersion (MSPD) and gas chromatography-mass spectrometry were used for the rapid determination of 18 plasticizers (phthalates and adipates), 7 polycyclic musks and 5 nitromusks, which makes a total of 30 targets, in both rinse-off and leave-on cosmetic formulations. The MSPD method was miniaturized and customized to avoid or minimize risks of phthalate contamination and to reduce residues and costs. The amount of sample and extraction solvent employed were 0.1g and 1mL, respectively. The procedure was optimized by means of an experimental design and under the optima conditions it showed satisfactory linearity, repeatability and intermediate precision. LOQs were, in general, in the low ngg(-1), and recoveries were quantitative for all the 18 plasticizers and the 12 fragrances. Twenty-six cosmetic products such as creams, emulsions, lotions, gels for the skin, bath and shower preparations, deodorants, hair-setting, hair-cleansing and hair-conditioning products, shaving products, and sunbathing products, were analyzed. Twenty-five out of thirty targets were detected in the samples. The most frequently found compounds were galaxolide and tonalide reaching concentrations above 0.1% (1000μgg(-1)), and diethyl phthalate (between 0.7 and 357μgg(-1)). The presence of banned substances (Regulation (EC) No. 1223/2009) such as dibutyl phthalate, diisobutyl phthalate, dimethoxyethyl phthalate, benzylbutyl phthalate, diethylhexyl phthalate, diisopentyl phthalate and dipentyl phthalate, musk ambrette and musk tibetene was confirmed in sixteen of the twenty-six personal care products (62%).
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, LR, ≥99%
Sigma-Aldrich
Ethyl acetate, AR, ≥99%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%