Merck
  • Synthesis of novel therapeutic agents for the treatment of multiple sclerosis: a brief overview.
Synthesis of novel therapeutic agents for the treatment of multiple sclerosis: a brief overview.
European journal of medicinal chemistry (2013-01-08)
Naveen Mulakayala, Pallavi Rao, Javed Iqbal, Rakeshwar Bandichhor, Srinivas Oruganti
ABSTRACT
Multiple sclerosis (MS) often results in chronic inflammatory and autoimmune disorders, and recent developments in understanding the disease pathogenesis has lead to newer therapeutic options for the treatment of the disease. The development of small molecule drugs with improved efficacy, better tolerability, and oral administration has received a new impetus with the discovery of newer classes of drugs. In this review, we have summarized the hitherto known synthetic strategies of fingolimod, laquinimod, cladribine, and teriflunomide reported in the literature which are the key small molecules and the first oral drug candidates for MS in various stages of clinical development or have been launched in the market.
Product Description
Cladribine for peak identification, European Pharmacopoeia (EP) Reference Standard