Merck
Steering supramolecular patterns by nucleobase-terminated molecules.
Chemical communications (Cambridge, England) (2012-12-04)
Cai Shen, Jacob R Cramer, Mikkel F Jacobsen, Lei Liu, Shuai Zhang, Mingdong Dong, Kurt V Gothelf, Flemming Besenbacher
ABSTRACT
Supramolecular patterns formed by adsorption from a liquid of nucleobase-terminated molecular rods on a graphite surface were investigated by scanning tunneling microscopy.
Sigma-Aldrich
Thymine, suitable for cell culture, BioReagent
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Thymine, Vetec, reagent grade, 99%