Merck
  • Triazolyl donor/acceptor chromophore decorated unnatural nucleosides and oligonucleotides with duplex stability comparable to that of a natural adenine/thymine pair.
Triazolyl donor/acceptor chromophore decorated unnatural nucleosides and oligonucleotides with duplex stability comparable to that of a natural adenine/thymine pair.
The Journal of organic chemistry (2012-11-23)
Subhendu Sekhar Bag, Sangita Talukdar, Katsuhiko Matsumoto, Rajen Kundu
ABSTRACT
We report the design and synthesis of triazolyl donor/acceptor unnatural nucleosides via click chemistry and studies on the duplex stabilization of DNA containing two such new nucleosides. The observed duplex stabilization among the self-pair/heteropair has been found to be comparable to that of a natural A/T pair. Our observations on the comparable duplex stabilization has been explained on the basis of possible π-π stacking and/or charge transfer interactions between the pairing partners. The evidence of ground-state charge transfer complexation came from the UV-vis spectra and the static quenching of fluorescence in a heteropair. We have also exploited one of our unnatural DNAs in stabilizing abasic DNA.
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Thymine, suitable for cell culture, BioReagent
Sigma-Aldrich
Thymine, Vetec, reagent grade, 99%